accessibility menu, dialog, popup

UserName

CH3OOOCH3ON

CAS RN 603-50-9

IUPAC Name: 4-{[4-(acetyloxy)phenyl](pyridin-2-yl)methyl}phenyl acetate
Synonyms: bicol fleet bisacodyl numark brand of bisacodyl schwarzhaupt brand of bisacodyl zetrax laxans ratiopharm bisacodyl preparation bisacodylum bisco-lax ysatfabrik brand of bisacodyl
Molecular Weight (g/mol): 361.4
Molecular Formula: C22H19NO4
InChi Key: KHOITXIGCFIULA-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(C=C1)C(C1=CC=C(OC(C)=O)C=C1)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (10)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (1)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (10)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)