missing translation for 'onlineSavingsMsg'
Learn More

Bisacodyl, 98+%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J61997.09

Quantity 10g

 View more versions of this product

Product Code. 15444469

  • 131.00 EUR / 10g
 Restricted Item
Estimated Shipment: 31-08-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in alcohol (slightly), ether (slightly), chloroform, and acetone. Insoluble in water.

Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

603-50-9
361.40
KHOITXIGCFIULA-UHFFFAOYSA-N
2391
CC(=O)OC1=CC=C(C=C1)C(C1=CC=C(OC(C)=O)C=C1)C1=CC=CC=N1
C22H19NO4
MFCD00038039
bisacodyl, dulcolax, bicol, brocalax, durolax, dulcolan, endokolat, fenilaxan, hillcolax, sanvacual
4-{[4-(acetyloxy)phenyl](pyridin-2-yl)methyl}phenyl acetate

Specifications

603-50-9
MFCD00038039
14,1242
Soluble in alcohol (slightly),ether (slightly),chloroform,and acetone. Insoluble in water.
CC(=O)OC1=CC=C(C=C1)C(C1=CC=C(OC(C)=O)C=C1)C1=CC=CC=N1
361.40
361.4
Bisacodyl
C22H19NO4
10g
bisacodyl, dulcolax, bicol, brocalax, durolax, dulcolan, endokolat, fenilaxan, hillcolax, sanvacual
KHOITXIGCFIULA-UHFFFAOYSA-N
4-{[4-(acetyloxy)phenyl](pyridin-2-yl)methyl}phenyl acetate
2391
≥98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.