missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress AMPPD

Product Code. 16449447
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449447 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16449447 Supplier MedChemExpress Supplier No. HY15906

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AMPPD, a 1,2-dioxo-cyclohexane derivative, is a biochemistry ultrasensitive alkaline phosphatase substrate.

Specifications

CAS 122341-56-4
Solubility H2O : 83.33 mg/mL (217.95 mM∣ultrasonic and warming and heat to 60°C)
Synonym Lumi-Phos Plus∣ Lumigen PPD∣ PPD
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Quantity 5 mg
Molecular Weight (g/mol) 382.34 g/mol
Product Type Biochemical Assay Reagents
Purity or Quality Grade 98.28%
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.