Learn More
Melford 4-Nitrophenyl-β-D-Galactopyranoside
Product Code. 30018616
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018616 5 g 5g
30018615 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30018616 Supplier Melford Supplier No. N810105.0
 Restricted Item
Estimated Shipment: 28-10-2026
Colorimetric and spectrophotoametric substrate for detection of B-D Galactosidase.
Colorimetric and spectrophotoametric substrate for detection of B-D Galactosidase.
Specifications
Quantity 5 g