Learn More
Melford 4-Nitrophenyl-β-D-Galactopyranoside
Product Code. 30018615
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 30018615
Brand: Melford N810101.0
Colorimetric and spectrophotoametric substrate for detection of B-D Galactosidase.
Colorimetric and spectrophotoametric substrate for detection of B-D Galactosidase.
Specifications
Quantity 1 g