Learn More
Melford 4-Methylumbelliferyl-α-L-arabinopyranoside
Product Code. 30018502
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018502 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30018502 Supplier Melford Supplier No. M641550.1
 Restricted Item
Estimated Shipment: 12-11-2026
Fluorescent substrate for α-L-Arabinosidase.
Fluorescent substrate for α-L-Arabinosidase. 4-Methylumbelliferone < 50ppm.