Learn More
Melford 4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-galactopyranoside
Product Code. 30018497
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018497 1 g 1g
30018496 100 mg 100mg
30018495 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30018497 Supplier Melford Supplier No. M639001.0
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
Fluorescent substrate for N-Acetyl-β-D-galactosaminidase.
Fluorescent substrate for N-Acetyl-β-D-galactosaminidase.
Specifications
Quantity 1 g