missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethanol, Sigma-Aldrich™

Product Code. 15844171
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Unit Size:
1 L
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15844171 1 L 1 L
15824171 5 mL 5mL
15834171 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15844171 Supplier Sigma Aldrich Supplier No. 8057401000
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-24-2

Specifications

Molecular Weight (g/mol) 78.13
Molecular Formula HSCH2CH2OH
Boiling Point 157°C
Density 1.114 g/mL at 25°C
Quantity 1 L
Synonym Thioethylene glycol, Thioglycol, β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol, β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol
Melting Point <-50°C
Format Standard
Product Type Mercaptoethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.