CAS RN 60-24-2
2-Mercaptoethanol, 99%, pure
CAS: 60-24-2 Molecular Formula: C2H6OS Molecular Weight (g/mol): 78.13 MDL Number: MFCD00004890 InChI Key: DGVVWUTYPXICAM-UHFFFAOYSA-N Synonym: 2-mercaptoethanol,mercaptoethanol,thioglycol,beta-mercaptoethanol,ethanol, 2-mercapto,2-thioethanol,2-hydroxy-1-ethanethiol,thioethylene glycol,2-hydroxyethanethiol,2-hydroxyethyl mercaptan PubChem CID: 1567 ChEBI: CHEBI:41218 SMILES: OCCS
Thermo Scientific™ Pierce™ 2-Mercaptoethanol
Pure, liquid (equals 14M), beta-mercaptoethanol (bME, 2BME, 2-ME, b-mer), CAS 60-24-2; thiol reducing agent for cleaving protein disulfide bonds (cystines cysteines).
2-Mercaptoethanol, Electrophoresis Grade, 98+%
CAS: 60-24-2 Molecular Formula: C2H6OS Molecular Weight (g/mol): 78.13 MDL Number: MFCD00004890 InChI Key: DGVVWUTYPXICAM-UHFFFAOYSA-N Synonym: 2-mercaptoethanol,mercaptoethanol,thioglycol,beta-mercaptoethanol,ethanol, 2-mercapto,2-thioethanol,2-hydroxy-1-ethanethiol,thioethylene glycol,2-hydroxyethanethiol,2-hydroxyethyl mercaptan PubChem CID: 1567 ChEBI: CHEBI:41218 SMILES: OCCS
2-Mercaptoethanol, 98+%
CAS: 60-24-2 Molecular Formula: C2H6OS Molecular Weight (g/mol): 78.13 MDL Number: MFCD00004890 InChI Key: DGVVWUTYPXICAM-UHFFFAOYSA-N Synonym: 2-mercaptoethanol,mercaptoethanol,thioglycol,beta-mercaptoethanol,ethanol, 2-mercapto,2-thioethanol,2-hydroxy-1-ethanethiol,thioethylene glycol,2-hydroxyethanethiol,2-hydroxyethyl mercaptan PubChem CID: 1567 ChEBI: CHEBI:41218 SMILES: OCCS
2-Mercaptoethanol Molecular Biology MP Biomedicals
2-Mercaptoethanol, ≥98%, MP Biomedicals™
CAS: 60-24-2 Molecular Formula: C2H6OS Molecular Weight (g/mol): 78.13 MDL Number: MFCD00004890 InChI Key: DGVVWUTYPXICAM-UHFFFAOYSA-N Synonym: 2-mercaptoethanol,mercaptoethanol,thioglycol,beta-mercaptoethanol,ethanol, 2-mercapto,2-thioethanol,2-hydroxy-1-ethanethiol,thioethylene glycol,2-hydroxyethanethiol,2-hydroxyethyl mercaptan PubChem CID: 1567 ChEBI: CHEBI:41218 SMILES: OCCS
2-Mercaptoethanol, >98%, MP Biomedicals™
2-Mercaptoethanol Cell Culture MP Biomedicals
CAS: 60-24-2 Molecular Formula: C2H6OS Molecular Weight (g/mol): 78.13 MDL Number: MFCD00004890 InChI Key: DGVVWUTYPXICAM-UHFFFAOYSA-N Synonym: 2-mercaptoethanol,mercaptoethanol,thioglycol,beta-mercaptoethanol,ethanol, 2-mercapto,2-thioethanol,2-hydroxy-1-ethanethiol,thioethylene glycol,2-hydroxyethanethiol,2-hydroxyethyl mercaptan PubChem CID: 1567 ChEBI: CHEBI:41218 IUPAC Name: 2-sulfanylethan-1-ol SMILES: OCCS
2-Mercaptoethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2-Mercaptoethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2-Mercaptoethanol, Sigma-Aldrich™
CAS: 60-24-2
2-Mercaptoethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.