CAS RN 99-80-9
CAS RN 99-80-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Elastopar, TRC
CAS: 99-80-9 Molecular Formula: C7 H7 N3 O2 Molecular Weight (g/mol): 165.149 Synonym: Aniline, N-methyl-N,p-dinitroso- (6CI,7CI,8CI),N-Methyl-N,4-dinitrosobenzenamine,Elastopar,Heat Pre,N,4-Dinitroso-N-methylaniline,N,4-Dinitroso-N-methylbenzenamine,N,p-Dinitroso-N-methylaniline,N-Methyl-N,4-dinitrosoaniline,Nitrosan K,Vulnoc DBM IUPAC Name: N-methyl-N-(4-nitrosophenyl)nitrous amide SMILES: CN(N=O)c1ccc(cc1)N=O