Learn More
Elastopar, TRC
Product Code. 30379581
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379581 100 mg 100mg
30378384 50 mg -
30373058 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30379581 Supplier Toronto Research Chemicals Supplier No. TRCE324550100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 99-80-9
Molecular Formula C7 H7 N3 O2
Molecular Weight (g/mol) 165.149
Synonym Aniline, N-methyl-N,p-dinitroso- (6CI,7CI,8CI), N-Methyl-N,4-dinitrosobenzenamine, Elastopar, Heat Pre, N,4-Dinitroso-N-methylaniline, N,4-Dinitroso-N-methylbenzenamine, N,p-Dinitroso-N-methylaniline, N-Methyl-N,4-dinitrosoaniline, Nitrosan K, Vulnoc DBM
IUPAC Name N-methyl-N-(4-nitrosophenyl)nitrous amide
SMILES CN(N=O)c1ccc(cc1)N=O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C7H7N3O2/c1-10(9-12)7-4-2-6(8-11)3-5-7/h2-5H,1H3
Formula Weight 165.0538
Chemical Name or Material N-Methyl-N,4-dinitrosobenzenamine