CAS RN 99-49-0
CAS RN 99-49-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Carvone, TRC
CAS: 99-49-0 Molecular Formula: C10 H14 O Molecular Weight (g/mol): 150.22 Synonym: p-Mentha-6,8-dien-2-one (8CI),2-Methyl-5-(1-methylethenyl)-2-cyclohexen-1-one,(±)-Carvone,2-Methyl-5-isopropenyl-2-cyclohexenone,5-Isopropenyl-2-methylcyclohex-2-enone,5-Isopropyl-2-methyl-2-cyclohexen-1-one,Carvone,DL-Carvone,Karvon,Limonen-6-one,NSC 6275,p-Mentha-1(6),8-dien-2-one IUPAC Name: 2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one SMILES: CC(=C)C1CC=C(C)C(=O)C1
2
Carvone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality