Learn More
Carvone, TRC
Product Code. 30135967
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135967 5 g 5g
30004117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135967 Supplier Toronto Research Chemicals Supplier No. TRCC1841955G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 99-49-0
Molecular Formula C10 H14 O
Molecular Weight (g/mol) 150.22
Synonym p-Mentha-6,8-dien-2-one (8CI), 2-Methyl-5-(1-methylethenyl)-2-cyclohexen-1-one, (±)-Carvone, 2-Methyl-5-isopropenyl-2-cyclohexenone, 5-Isopropenyl-2-methylcyclohex-2-enone, 5-Isopropyl-2-methyl-2-cyclohexen-1-one, Carvone, DL-Carvone, Karvon, Limonen-6-one, NSC 6275, p-Mentha-1(6),8-dien-2-one
IUPAC Name 2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
SMILES CC(=C)C1CC=C(C)C(=O)C1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9H,1,5-6H2,2-3H3
Formula Weight 150.1045
Chemical Name or Material Carvone