Learn More
Carvone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30326816
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326816 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30326816 Supplier Dr. Ehrenstorfer Supplier No. DREA11052000AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Carvone
CAS 99-49-0
Analyte or Component Names Carvone
Shipping Condition Room Temperature
Molecular Formula C10 H14 O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9H,1,5-6H2,2-3H3
SMILES CC(=C)C1CC=C(C)C(=O)C1
IUPAC Name 2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 150.22
Quantity 1 mL
Formula Weight 150.1045 g/mol