accessibility menu, dialog, popup

UserName

CAS RN 950649-05-5

IUPAC Name: 4-[2-(trifluoromethoxy)phenoxy]piperidine hydrochloride
Synonyms: 4-[2-(trifluoromethoxy)phenoxy]piperidine hydrochloride
Molecular Weight (g/mol): 297.7
Molecular Formula: C12H15ClF3NO2
InChi Key: DWPFMBFQDUMIEP-UHFFFAOYSA-N
SMILES: Cl.FC(F)(F)OC1=CC=CC=C1OC1CCNCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results