missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Trifluoromethoxy)phenoxy]piperidine hydrochloride, Thermo Scientific™

Product Code. 15499205
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499205 250 mg 250mg
15489205 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499205 Supplier Thermo Scientific Alfa Aesar Supplier No. H51178.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 950649-05-5
Molecular Formula C12H15ClF3NO2
Molecular Weight (g/mol) 297.702
MDL Number MFCD11870723
InChI Key DWPFMBFQDUMIEP-UHFFFAOYSA-N
Synonym 4-2-trifluoromethoxy phenoxy piperidine hydrochloride, 4-2-trifluoromethoxy-phenoxy-piperidine hydrochloride, 4-2-trifluoromethoxy phenoxy piperidine,hydrochloride
PubChem CID 53400828
IUPAC Name 4-[2-(trifluoromethoxy)phenoxy]piperidine;hydrochloride
SMILES C1CNCCC1OC2=CC=CC=C2OC(F)(F)F.Cl

Specifications

Quantity 250 mg
Beilstein 15564237
Formula Weight 297.71
Chemical Name or Material 4-[2-(Trifluoromethoxy)phenoxy]piperidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.