accessibility menu, dialog, popup

UserName

H3CNONOSOOHOOHH2N

CAS RN 936095-50-0

IUPAC Name: 3-{[3-(2-amino-2-carboxyethyl)-5-methyl-2,6-dioxo-1,3-diazinan-1-yl]methyl}-5-phenylthiophene-2-carboxylic acid
Synonyms: 3-{[3-(2-amino-2-carboxyethyl)-5-methyl-2,6-dioxo-1,3-diazinan-1-yl]methyl}-5-phenylthiophene-2-carboxylic acid
Molecular Weight (g/mol): 431.46
Molecular Formula: C20H21N3O6S
InChi Key: OAAMYRWMIRGBQF-UHFFFAOYNA-N
SMILES: CC1CN(CC(N)C(O)=O)C(=O)N(CC2=C(SC(=C2)C2=CC=CC=C2)C(O)=O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results