accessibility menu, dialog, popup

UserName

CH3HNNOH3C

CAS RN 916792-58-0

IUPAC Name: 1-[3-(ethylamino)pyrrolidin-1-yl]ethan-1-one
Synonyms: 1-[3-(ethylamino)pyrrolidin-1-yl]ethanone
Molecular Weight (g/mol): 156.23
Molecular Formula: C8H16N2O
InChi Key: GILRDSYIJNXDLB-UHFFFAOYNA-N
SMILES: CCNC1CCN(C1)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results