Learn More
Alfa Aesar™ 1-Acetyl-3-ethylaminopyrrolidine, 99%
Product Code. 11499780
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499780 250 mg 250mg
11409790 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11499780 Supplier Alfa Aesar™ Supplier No. H51001.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 916792-58-0
Molecular Formula C8H16N2O
Molecular Weight (g/mol) 156.229
MDL Number MFCD08061108
InChI Key GILRDSYIJNXDLB-UHFFFAOYSA-N
Synonym 1-acetyl-3-ethylaminopyrrolidine, 1-acetyl-n-ethylpyrrolidin-3-amine, 1-3-ethylamino pyrrolidin-1-yl ethan-1-one, 1-3-ethylamino pyrrolidin-1-yl ethanone, acmc-20aoyw, 1-3-ethylamino-pyrrolidin-1-yl-ethanone
PubChem CID 45934254
IUPAC Name 1-[3-(ethylamino)pyrrolidin-1-yl]ethanone
SMILES CCNC1CCN(C1)C(=O)C
Specifications
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 156.23
Percent Purity 99%
Chemical Name or Material 1-Acetyl-3-ethylaminopyrrolidine