accessibility menu, dialog, popup

UserName

CAS RN 9007-49-2

A(a)

CAS RN 9007-49-2

IUPAC Name: [(5-amino-2-{[({[5-amino-2-(hydroxymethyl)oxolan-3-yl]oxy}(hydroxy)phosphoryl)oxy]methyl}oxolan-3-yl)oxy][(5-amino-3-hydroxyoxolan-2-yl)methoxy]phosphinic acid
Synonyms: DNA
Molecular Weight (g/mol): 523.37
Molecular Formula: C15H31N3O13P2
InChi Key: AWBASQCACWFTGD-UHFFFAOYNA-N
SMILES: NC1CC(O)C(COP(O)(=O)OC2CC(N)OC2COP(O)(=O)OC2CC(N)OC2CO)O1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results