missing translation for 'onlineSavingsMsg'
Learn More

Deoxyribonucleic acid sodium salt, MP Biomedicals™

Product Code. 11454342 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11454342

Brand: MP Biomedicals 0216009801

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 9007-49-2
Color White
Molecular Formula C15H31N3O13P2
MDL Number MFCD00130921
Quantity 1 g
Synonym nucleic acid, na salt yeast, deoxyribonucleic acid herring sperm , not higly polimerized, 5-amino-2-5-amino-2-hydroxymethyl oxolan-3-yl oxy hydroxy phosphoryl oxy methyl oxolan-3-yl oxy 5-amino-3-hydroxyoxolan-2-yl methoxyphosphinic acid
InChI Key AWBASQCACWFTGD-UHFFFAOYNA-N
SMILES *
IUPAC Name Deoxyribonucleic acid
Molecular Weight (g/mol) 0.00
PubChem CID 44135672
Physical Form Powder
Chemical Name or Material Deoxyribonucleic acid sodium salt
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.