missing translation for 'onlineSavingsMsg'
Learn More

Deoxyribonucleic Acid Sodium Salt, MP Biomedicals™

Product Code. 11464342 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464342 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11464342 Supplier MP Biomedicals Supplier No. 0216009805
  • ONLINE EXCLUSIVE PRICE
    687.00 EUR
    654.15 EUR / 5g
    Save 32.85 EUR
    4% Off
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains information in chemical code to direct the development of the cell according to its inheritance. Deoxyribonucleic Acid Sodium Salt, MP Biomedicals (DNA) is a polynucleotide and an essential component of chromosomes in cell nuclei. It is the carri

Deoxyribonucleic acid sodium salt has been used in a study to evaluate the cytotoxicity of enzyme inhibitors and absorption enhancers in Caco-2 cells for oral delivery of salmon calcitonin. Deoxyribonucleic acid sodium salt has also been used in a study to develop a simple method for extracting DNA from Cryptosporidium oocysts. It also acts as a template in the formation of ribonucleic acids, which play a fundamental role in the synthesis of proteins in the cell.

  • The purine and pyrimidine bases of the nucleosides are primarily adenine, guanine, cytosine and thymine
  • The sugar is D-2-deoxyribose
  • The nucleosides are linked together by phosphates in diester linkage from the 3'-hydroxyl of one sugar to the 5'-hydroxyl of the next
  • The repeating sugar-phosphate linkage forms the backbone of the single polynucleotide strand, which is the primary structure of DNA
  • Chemical analysis of DNA from different species show that the purine content is equal to the pyrimidine content; adenine content equal to thymine; guanine equal to cytosine

Specifications

CAS 9007-49-2
Molecular Formula C15H31N3O13P2
MDL Number MFCD00130921
Quantity 5 g
Synonym nucleic acid, na salt yeast, deoxyribonucleic acid herring sperm , not higly polimerized, 5-amino-2-5-amino-2-hydroxymethyl oxolan-3-yl oxy hydroxy phosphoryl oxy methyl oxolan-3-yl oxy 5-amino-3-hydroxyoxolan-2-yl methoxyphosphinic acid
Solubility Information Most DNA is soluble in water or aqueous buffers. Slight heating may help. The addition of SDS or similar detergent may help solubilize DNA; however, detergents tend to denature the DNA into shorter strands.
InChI Key AWBASQCACWFTGD-UHFFFAOYNA-N
SMILES *
IUPAC Name Deoxyribonucleic acid
Molecular Weight (g/mol) 0.00
PubChem CID 44135672
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.