accessibility menu, dialog, popup

UserName

CAS RN 86-96-4

OHNONH

CAS RN 86-96-4

IUPAC Name: 1,2,3,4-tetrahydroquinazoline-2,4-dione
Synonyms: 1,3-dihydroquinazoline-2,4-dione
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: SDQJTWBNWQABLE-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results