missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzouracil, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Benzouracil |
| CAS | 86-96-4 |
| Molecular Formula | C8H6N2O2 |
| Quantity | 5 g |
| Synonym | 1,2,3,4-Tetrahydro-2,4-dioxoquinazoline, 1,2,3,4-Tetrahydroquinazoline-2,4-dione, 2,4-Dihydroxyquinazoline, 2,4-Dioxotetrahydroquinazoline, 2,4-Quinazolinediol, Benzoyleneurea, NSC 2108, Quinazoline-2,4-dione, Quinazolinedione, y-Thymine, yT |
| InChI Formula | InChI=1S/C8H6N2O2/c11-7-5-3-1-2-4-6(5)9-8(12)10-7/h1-4H,(H2,9,10,11,12) |
| SMILES | O=C1NC(=O)c2ccccc2N1 |
| IUPAC Name | 1H-quinazoline-2,4-dione |
| Molecular Weight (g/mol) | 162.15 |
| Formula Weight | 162.0429 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction