Learn More
Benzoyleneurea, 98%
Product Code. 11342047
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11342047
Brand: Thermo Scientific Alfa Aesar L07763.14
Request Bulk or Custom Format
CAS: 86-96-4 | C8H6N2O2 | 162.148 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 86-96-4
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00006699
InChI Key SDQJTWBNWQABLE-UHFFFAOYSA-N
Synonym benzoyleneurea, quinazoline-2,4 1h,3h-dione, 2,4 1h,3h-quinazolinedione, quinazoline-2,4-diol, 2,4-dihydroxyquinazoline, quinazolinedione, benzouracil, quinazoline-2,4-dione, urea, benzoylene, 1,2,3,4-tetrahydroquinazoline-2,4-dione
PubChem CID 64048
IUPAC Name 1H-quinazoline-2,4-dione
SMILES C1=CC=C2C(=C1)C(=O)NC(=O)N2
Specifications
Melting Point >300°C
Quantity 25 g
Beilstein 383776
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material Benzoyleneurea