accessibility menu, dialog, popup

UserName

NClFF

CAS RN 851179-00-5

IUPAC Name: 3-chloro-2,5-difluoropyridine
Synonyms: 3-chloro-2,5-difluoropyridine
Molecular Weight (g/mol): 149.52
Molecular Formula: C5H2ClF2N
InChi Key: ATKDHGGWYZLPPJ-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=C(F)N=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)