missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-2,5-difluoropyridine, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-2,5-difluoropyridine |
| CAS | 851179-00-5 |
| Molecular Formula | C5H2ClF2N |
| Quantity | 100 mg |
| Synonym | 3-Chloro-2,5-difluoropyridine (ACI) |
| InChI Formula | InChI=1S/C5H2ClF2N/c6-4-1-3(7)2-9-5(4)8/h1-2H |
| SMILES | ClC(C=C(F)C=N1)=C1F |
| IUPAC Name | 3-chloro-2,5-difluoropyridine |
| Molecular Weight (g/mol) | 149.53 |
| Formula Weight | 148.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction