missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,5-difluoropyridine, TRC

Product Code. 30349336
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30349336 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349336 Supplier Toronto Research Chemicals Supplier No. TRCC368285250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Chloro-2,5-difluoropyridine
CAS 851179-00-5
Molecular Formula C5H2ClF2N
Quantity 250 mg
Synonym 3-Chloro-2,5-difluoropyridine (ACI)
InChI Formula InChI=1S/C5H2ClF2N/c6-4-1-3(7)2-9-5(4)8/h1-2H
SMILES ClC(C=C(F)C=N1)=C1F
IUPAC Name 3-chloro-2,5-difluoropyridine
Molecular Weight (g/mol) 149.53
Formula Weight 148.98
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.