accessibility menu, dialog, popup

UserName

BOHNNNHNNOHOHHCl

CAS RN 850567-38-3

IUPAC Name: {3-[(2H-1,2,3,4-tetrazol-5-yl)carbamoyl]phenyl}boronic acid hydrochloride
Synonyms: 3-[(2H-1,2,3,4-tetrazol-5-yl)carbamoyl]phenylboronic acid hydrochloride
Molecular Weight (g/mol): 269.45
Molecular Formula: C8H9BClN5O3
InChi Key: PWSGJXZLSYWEAJ-UHFFFAOYSA-N
SMILES: Cl.OB(O)C1=CC(=CC=C1)C(=O)NC1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results