missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Tetrazol-5-ylcarbamoyl)benzeneboronic acid hydrochloride, 97%, Thermo Scientific™

Product Code. 15499885
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499885 1 g 1g
15409895 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499885 Supplier Thermo Scientific Alfa Aesar Supplier No. H52537.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850567-38-3
Molecular Formula C8H9BClN5O3
Molecular Weight (g/mol) 269.452
MDL Number MFCD06659888
InChI Key PWSGJXZLSYWEAJ-UHFFFAOYSA-N
Synonym 3-1h-tetrazol-5-yl carbamoyl phenyl boronic acid hydrochloride, 3-1h-tetrazol-5-ylcarbamoyl benzeneboronic acid hydrochloride, 3-1h-tetrazol-5-yl-carbamoyl phenylboronic acid, hcl, 3-1h-tetrazol-5-yl-carbamoyl benzeneboronic acid, hcl, 3-1h-1,2,3,4-tetrazol-5-yl carbamoyl phenylboronic acid hydrochloride, 3-1h-tetrazol-5-yl-carbamoyl phenylboronic acid hcl, 3-1h-tetrazol-5-yl-carbamoyl phenylboronic acid hydrochloride, 3-1h-tetrazol-5-ylaminocarbonyl phenylboronic acid hydrochloride, 3-2h-tetrazol-5-yl carbamoyl phenyl boronic acid-hydrogen chloride 1/1, 3-2h-1,2,3,4-tetrazol-5-yl carbamoyl phenylboronic acid hydrochloride
PubChem CID 44119544
IUPAC Name [3-(2H-tetrazol-5-ylcarbamoyl)phenyl]boronic acid;hydrochloride
SMILES B(C1=CC(=CC=C1)C(=O)NC2=NNN=N2)(O)O.Cl

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 269.45
Percent Purity 97%
Chemical Name or Material 3-(1H-Tetrazol-5-ylcarbamoyl)benzeneboronic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.