CAS RN 848318-25-2
CAS RN 848318-25-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
SSR128129E, MedChemExpress
MedChemExpress SSR128129E is an orally available and allosteric FGFR inhibitor with an IC50 of 1.9 μM for FGFR1.