Learn More
SSR128129E, TRC
Specifications
Specifications
| Chemical Name or Material | SSR128129E |
| CAS | 848318-25-2 |
| Molecular Formula | C18H15N2NaO4 |
| Quantity | 100 mg |
| Synonym | 2-Amino-5-[(1-methoxy-2-methyl-3-indolizinyl)carbonyl]-benzoic Acid Sodium Salt, 2-Amino-5-[(1-methoxy-2-methyl-3-indolizinyl)carbonyl]-benzoic Acid Monosodium Salt, SSR 128129E, Sodium 2-Amino-5-[(1-methoxy-2-methylindolizin-3-yl)carbonyl]benzoate |
| InChI Formula | InChI=1S/C18H16N2O4.Na/c1-10-15(20-8-4-3-5-14(20)17(10)24-2)16(21)11-6-7-13(19)12(9-11)18(22)23;/h3-9H,19H2,1-2H3,(H,22,23);/q;+1/p-1 |
| SMILES | O=C(C1=CC(C(O[Na])=O)=C(N)C=C1)C(N2C=CC=CC2=C3OC)=C3C |
| Molecular Weight (g/mol) | 346.31 |
| Formula Weight | 346.093 |