Learn More
SSR128129E, MedChemExpress
Product Code. 16482747
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482747 50 mg 50mg
16446668 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16482747 Supplier Medchem Express Supplier No. HY15599
MedChemExpress SSR128129E is an orally available and allosteric FGFR inhibitor with an IC50 of 1.9 μM for FGFR1.
SSR128129E is an orally available and allosteric FGFR inhibitor with an IC50 of 1.9 μM for FGFR1.
Specifications
Chemical Name or Material SSR128129E
Physical Form Solid
CAS 848318-25-2
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H15N2NaO4
For Use With (Application) Cancer-Kinase/protease
Synonym SSR
Solubility Information DMSO : ≥ 54 mg/mL (155.93 mM)
SMILES O=C(O[Na])C1=CC(C(C2=C(C)C(OC)=C3C=CC=CN23)=O)=CC=C1N
Molecular Weight (g/mol) 346.31
Formula Weight 346.31
Percent Purity 99.86%
Grade Research