accessibility menu, dialog, popup

UserName

CAS RN 833-66-9

KHOSOOO

CAS RN 833-66-9

IUPAC Name: potassium 6-hydroxynaphthalene-2-sulfonate
Synonyms: potassium 6-hydroxynaphthalene-2-sulfonate
Molecular Weight (g/mol): 262.32
Molecular Formula: C10H7KO4S
InChi Key: PZWWSVPMGHDZNJ-UHFFFAOYSA-M
SMILES: [K+].OC1=CC=C2C=C(C=CC2=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)