missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxynaphthalene-2-sulfonic Acid Potassium Salt, TRC

Product Code. 30136182
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136182 100 mg 100mg
30135946 50 mg 50mg
30012099 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136182 Supplier Toronto Research Chemicals Supplier No. TRCH951188100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 833-66-9
Molecular Formula C10 H7 O4 S . K
Molecular Weight (g/mol) 262.3235
IUPAC Name potassium;6-hydroxynaphthalene-2-sulfonate
SMILES [K+].Oc1ccc2cc(ccc2c1)S(=O)(=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H8O4S.K/c11-9-3-1-8-6-10(15(12,13)14)4-2-7(8)5-9;/h1-6,11H,(H,12,13,14);/q;+1/p-1
Formula Weight 261.9702
Chemical Name or Material 6-Hydroxynaphthalene-2-sulfonic Acid Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.