accessibility menu, dialog, popup

UserName

CAS RN 83-73-8

OHIIN

CAS RN 83-73-8

IUPAC Name: 5,7-diiodoquinolin-8-ol
Synonyms: iodoquinol diodoxyquinoleine iodoquinol usp entero-diyod caswell no. 354 diodoquin diiodohydroxyquinolinum diodoxylin di-quinol searle brand of iodoquinol
Molecular Weight (g/mol): 396.95
Molecular Formula: C9H5I2NO
InChi Key: UXZFQZANDVDGMM-UHFFFAOYSA-N
SMILES: OC1=C2N=CC=CC2=C(I)C=C1I
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results