Learn More
Iodoquinol (5,7-Diiodoquinolin-8-ol), LoGiCal
Description
Iodoquinol (5,7-Diiodoquinolin-8-ol)
Specifications
Specifications
| Chemical Name or Material | Iodoquinol (5,7-Diiodoquinolin-8-ol) |
| CAS | 83-73-8 |
| Analyte or Component Names | Iodoquinol (5,7-Diiodoquinolin-8-ol) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H5 I2 N O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 5,7-Diiodoquinolin-8-ol, Clioquinol Imp. C (EP), 5,7-Diiodo-8-hydroxyquinoline, 5,7-Diiodo-8-quinolinol, 5,7-Diiodooxine, 8-Hydroxy-5,7-diiodoquinoline, Di-Quinol, Diamoebin, Diiodohydroxyquin, Diiodohydroxyquinoline, Diiodoquin, Diiodoquinol, Dinoleine, Diodohydroxyquin, Diodoquin, Diodoxylin, Direxiode, Disoquin, Dyodin, Embequin, Enterodiamoebin, Enterosept, Floraquin, Iodoquinol, Ioquin, Lanodoxin, Moebiquin, NSC 74939, NSC 8704, Quinadome, Rafamebin, SS 578, Searlequin, Searlewuin, Sebaquin, Stanquinate, Yodoxin, Zoaquin, Clioquinol Impurity C |
| InChI Formula | InChI=1 S/C9H5I2NO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13 H |
| SMILES | Oc1c(I)cc(I)c2cccnc12 |
| IUPAC Name | 5,7-diiodoquinolin-8-ol |
| Show More |