missing translation for 'onlineSavingsMsg'
Learn More

Iodoquinol, TRC

Product Code. 17977454
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17977454 2.5 g 2.5g
17928114 100 mg 100mg
17908424 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17977454 Supplier Toronto Research Chemicals Supplier No. TRCI7284002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-73-8
Molecular Formula C9 H5 I2 N O
Molecular Weight (g/mol) 396.95
Synonym 5,7-Diiodoquinolin-8-ol, Clioquinol Imp. C (EP), 5,7-Diiodo-8-hydroxyquinoline, 5,7-Diiodo-8-quinolinol, 5,7-Diiodooxine, 8-Hydroxy-5,7-diiodoquinoline, Di-Quinol, Diamoebin, Diiodohydroxyquin, Diiodohydroxyquinoline, Diiodoquin, Diiodoquinol, Dinoleine, Diodohydroxyquin, Diodoquin, Diodoxylin, Direxiode, Disoquin, Dyodin, Embequin, Enterodiamoebin, Enterosept, Floraquin, Iodoquinol, Ioquin, Lanodoxin, Moebiquin, NSC 74939, NSC 8704, Quinadome, Rafamebin, SS 578, Searlequin, Searlewuin, Sebaquin, Stanquinate, Yodoxin, Zoaquin
IUPAC Name 5,7-diiodoquinolin-8-ol
SMILES Oc1c(I)cc(I)c2cccnc12

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C9H5I2NO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13H
Formula Weight 396.846
Chemical Name or Material Iodoquinol (5,7-Diiodoquinolin-8-ol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.