accessibility menu, dialog, popup

UserName

CAS RN 81280-12-8

CH3H3COHH3C(E)

CAS RN 81280-12-8

IUPAC Name: (4E)-4-methylhept-4-en-3-ol
Synonyms: (4E)-4-methylhept-4-en-3-ol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: CVBRKUMLJLIZGY-VOTSOKGWNA-N
SMILES: CC\C=C(/C)C(O)CC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results