CAS RN 79722-21-7
CAS RN 79722-21-7
IUPAC Name:
tert-butyl N-(benzyloxy)carbamate
Synonyms:
tert-butyl N-(benzyloxy)carbamate
Molecular Weight (g/mol):
223.27
Molecular Formula:
C12H17NO3
InChi Key:
MZNBNPWFHGWAGH-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NOCC1=CC=CC=C1
1
–
3
of
3
results
1
tert-Butyl N-(Benzyloxy)carbamate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
tert-Butyl N-(Benzyloxy)carbamate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
tert-Butyl N-(Benzyloxy)carbamate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.