missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-(Benzyloxy)carbamate, TRC

Product Code. 30331009
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331009 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331009 Supplier Toronto Research Chemicals Supplier No. TRCB6928905G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl N-(Benzyloxy)carbamate
CAS 79722-21-7
Molecular Formula C12H17NO3
Quantity 5 g
Synonym tert-Butyl Benzyloxycarbamate, (Phenylmethoxy)carbamic Acid 1,1-Dimethylethyl Ester, 1,1-Dimethylethyl [(Phenylmethyl)oxy]carbamate, N-(tert-Butoxycarbonyl)-O-benzylhydroxylamine, N-Boc-O-benzylhydroxylamine, tert-Butyl (Benzyloxy)carbamate, tert-Butyl N-Benzyloxycarbamate
InChI Formula InChI=1S/C12H17NO3/c1-12(2,3)16-11(14)13-15-9-10-7-5-4-6-8-10/h4-8H,9H2,1-3H3,(H,13,14)
SMILES O=C(OC(C)(C)C)NOCC1=CC=CC=C1
IUPAC Name tert-butyl (benzyloxy)carbamate
Molecular Weight (g/mol) 223.27
Formula Weight 223.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.