accessibility menu, dialog, popup

UserName

CAS RN 79-81-2

IUPAC Name: (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl hexadecanoate
Synonyms: vitamin a palmitate vitamin a palmitate preparation retinol-15-3h arovit retinyl palmitate trans-retinyl palmitate optovit A optovit-A vi-dom-A lutavit A 500 plus
Molecular Weight (g/mol): 524.87
Molecular Formula: C36H60O2
InChi Key: VYGQUTWHTHXGQB-FFHKNEKCSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
Molecular Weight (g/mol) 
  • (4)
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (2)
  • (5)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (6)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (5)
  • (1)