missing translation for 'onlineSavingsMsg'
Learn More

Vitamin A palmitate, MP Biomedicals™

Product Code. 11457481 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457481 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11457481 Supplier MP Biomedicals Supplier No. 0210326210
  • ONLINE EXCLUSIVE PRICE
    73.85 EUR
    70.35 EUR / 10g
    Save 3.50 EUR
    4% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 79-81-2
Molecular Formula C36H60O2
Quantity 10 g
Synonym Retinyl plamitate
InChI Key VYGQUTWHTHXGQB-FFHKNEKCSA-N
SMILES CCCCCCCCCCCCCCCC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl hexadecanoate
Molecular Weight (g/mol) 524.87
Formula Weight 524.9
Physical Form Powder
Chemical Name or Material Vitamin A palmitate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.