missing translation for 'onlineSavingsMsg'
Learn More

Vitamin A palmitate, MP Biomedicals™

Product Code. 11467481 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467481 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11467481 Supplier MP Biomedicals Supplier No. 0210326280
  • ONLINE EXCLUSIVE PRICE
    106.00 EUR
    100.80 EUR / 100g
    Save 5.20 EUR
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 79-81-2
Color Yellow to Orange
Molecular Formula C36H60O2
Quantity 100 g
Synonym retinyl palmitate, vitamin a palmitate, retinol palmitate, retinol, hexadecanoate, arovit, all-trans-retinyl palmitate, optovit-a, retinyl hexadecanoate, aquapalm, dispatabs tabs
InChI Key VYGQUTWHTHXGQB-FFHKNEKCSA-N
SMILES CCCCCCCCCCCCCCCC(=O)OCC=C(C)C=CC=C(C)C=CC1=C(CCCC1(C)C)C
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] hexadecanoate
Molecular Weight (g/mol) 524.874
PubChem CID 5280531
ChEBI CHEBI:17616
Formula Weight 524.9
Physical Form Powder
Chemical Name or Material Vitamin A palmitate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.