accessibility menu, dialog, popup

UserName

H3CNHHNSSNCH3CH3NNOO(E)

CAS RN 76963-41-2

IUPAC Name: dimethyl[(4-{[(2-{[(1E)-1-(methylamino)-2-nitroethenyl]amino}ethyl)sulfanyl]methyl}-1,3-thiazol-2-yl)methyl]amine
Synonyms: nizatidine cronizat niatidine galitidin tazac zinga panaxid nizatidina antizid zanizal
Molecular Weight (g/mol): 331.45
Molecular Formula: C12H21N5O2S2
InChi Key: SGXXNSQHWDMGGP-IZZDOVSWSA-N
SMILES: CN\C(NCCSCC1=CSC(CN(C)C)=N1)=C/[N+]([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results