missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nizatidine, MedChemExpress
Description
Nizatidine is a potent and orally active histamine H2 receptor antagonist, can be used for the research of stomach and intestines ulcers. Nizatidine works by decreasing the secretion of gastric acid the stomach makes and prevent ulcers from coming back after they have healed in animal models.
Specifications
Specifications
| Chemical Name or Material | Nizatidine |
| Physical Form | Solid |
| CAS | 76963-41-2 |
| Color | Light Yellow |
| Quantity | 5 g |
| Purity Grade Notes | Research |
| Molecular Formula | C12H21N5O2S2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : ≥ 50 mg/mL (150.85 mM) ∣H2O : 20 mg/mL (60.34 mM; Need ultrasonic) |
| SMILES | CN/C(NCCSCC1=CSC(CN(C)C)=N1)=C\[N+]([O-])=O |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?