missing translation for 'onlineSavingsMsg'
Learn More

Nizatidine, TRC

Product Code. 30465489
Change view
Click to view available options
Quantity:
50 mg
1 g
25 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465489 1 g 1g
30478997 25 g -
30480071 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30465489 Supplier Toronto Research Chemicals Supplier No. TRCN5985001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76963-41-2
Molecular Formula C12 H21 N5 O2 S2
Molecular Weight (g/mol) 331.46
Synonym Nizatidine, N'-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N-methyl-2-nitro-1,1-ethenediamine, N-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N'-methyl-2-nitro-1,1-ethenediamine, Acinon, Axid, Bonacid, Calmaxid, Cronizat, Distaxid, Gastrax, LY 139037, Naxidine, Nizax, Nizaxid, Tazac, ZL 101, Zanizal
IUPAC Name 1-N'-[2-[[2-[(dimethylamino)methyl]-1,3-thiazol-4-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine
SMILES CNC(=C[N+](=O)[O-])NCCSCc1csc(CN(C)C)n1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H21N5O2S2/c1-13-11(6-17(18)19)14-4-5-20-8-10-9-21-12(15-10)7-16(2)3/h6,9,13-14H,4-5,7-8H2,1-3H3
Formula Weight 331.1137
Chemical Name or Material Nizatidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.