CAS RN 73-32-5
Thermo Scientific Chemicals L-Isoleucine, 99%
CAS: 73-32-5 Molecular Formula: C6H13NO2 Molecular Weight (g/mol): 131.18 MDL Number: MFCD00064222 MFCD00004268 InChI Key: AGPKZVBTJJNPAG-WHFBIAKZSA-N Synonym: l-isoleucine,isoleucine,2s,3s-2-amino-3-methylpentanoic acid,s-isoleucine,s,s-isoleucine,2s,3s-isoleucine,2-amino-3-methylvaleric acid,erythro-l-isoleucine,l-+-isoleucine,l-ile PubChem CID: 6306 ChEBI: CHEBI:17191 IUPAC Name: (2S,3S)-2-amino-3-methylpentanoic acid SMILES: CC[C@H](C)[C@H](N)C(O)=O
Thermo Scientific Chemicals L-Isoleucine, Cell Culture Reagent
CAS: 73-32-5 Molecular Formula: C6H13NO2 Molecular Weight (g/mol): 131.18 MDL Number: MFCD00064222 MFCD00004268 InChI Key: AGPKZVBTJJNPAG-WHFBIAKZSA-N Synonym: l-isoleucine,isoleucine,2s,3s-2-amino-3-methylpentanoic acid,s-isoleucine,s,s-isoleucine,2s,3s-isoleucine,2-amino-3-methylvaleric acid,erythro-l-isoleucine,l-+-isoleucine,l-ile PubChem CID: 6306 ChEBI: CHEBI:17191 IUPAC Name: (2S,3S)-2-amino-3-methylpentanoic acid SMILES: CC[C@H](C)[C@H](N)C(O)=O
Thermo Scientific Chemicals L-Isoleucine, 99%
CAS: 73-32-5 Molecular Formula: C6H13NO2 Molecular Weight (g/mol): 131.18 MDL Number: MFCD00064222 MFCD00004268 InChI Key: AGPKZVBTJJNPAG-WHFBIAKZSA-N Synonym: l-isoleucine,isoleucine,2s,3s-2-amino-3-methylpentanoic acid,s-isoleucine,s,s-isoleucine,2s,3s-isoleucine,2-amino-3-methylvaleric acid,erythro-l-isoleucine,l-+-isoleucine,l-ile PubChem CID: 6306 ChEBI: CHEBI:17191 IUPAC Name: (2S,3S)-2-amino-3-methylpentanoic acid SMILES: CC[C@H](C)[C@H](N)C(O)=O
L-Isoleucine, 99%, MP Biomedicals
CAS: 73-32-5 Molecular Formula: C6H13NO2 Molecular Weight (g/mol): 131.18 MDL Number: MFCD00064222 MFCD00004268 InChI Key: AGPKZVBTJJNPAG-WHFBIAKZSA-N Synonym: l-isoleucine,isoleucine,2s,3s-2-amino-3-methylpentanoic acid,s-isoleucine,s,s-isoleucine,2s,3s-isoleucine,2-amino-3-methylvaleric acid,erythro-l-isoleucine,l-+-isoleucine,l-ile PubChem CID: 6306 ChEBI: CHEBI:17191 IUPAC Name: (2S,3S)-2-amino-3-methylpentanoic acid SMILES: CC[C@H](C)[C@H](N)C(O)=O
L-Isoleucine, TRC
CAS: 73-32-5 Molecular Formula: C6 H13 N O2 Molecular Weight (g/mol): 131.17 Synonym: Isoleucine,L-Isoleucine,Isoleucine, L- (8CI),(2S,3S)-2-Amino-3-methylpentanoic acid,(2S,3S)-α-Amino-β-methyl-n-valeric acid,(2S,3S)-α-Amino-β-methylvaleric acid,(S)-Isoleucine,(S,S)-Isoleucine,2-Amino-3-methylvaleric acid,2S,3S-Isoleucine,Isoleucine, (2S,3ξ)-,L-(+)-Isoleucine,L-Ile,L-Norvaline, 3-methyl-, erythro-,NSC 46708,Pentanoic acid, 2-amino-3-methyl-, (2S,3S)-,[S-(R*,R*)]-2-Amino-3-methylpentanoic acid,erythro-L-Isoleucine IUPAC Name: (2S,3S)-2-amino-3-methylpentanoic acid SMILES: CC[C@H](C)[C@H](N)C(=O)O
(S)-(+)-Isoleucine, Sigma-Aldrich™
CAS: 73-32-5 Grade: Research Molecular Weight (g/mol): 131.17
L-Isoleucine, Multi-Compendial, U.S.P., J.T. Baker™
CAS: 73-32-5 Molecular Formula: C6H13NO2 Molecular Weight (g/mol): 131.18 MDL Number: MFCD00064222 MFCD00004268 InChI Key: AGPKZVBTJJNPAG-WHFBIAKZSA-N Synonym: l-isoleucine,isoleucine,2s,3s-2-amino-3-methylpentanoic acid,s-isoleucine,s,s-isoleucine,2s,3s-isoleucine,2-amino-3-methylvaleric acid,erythro-l-isoleucine,l-+-isoleucine,l-ile PubChem CID: 6306 ChEBI: CHEBI:17191 IUPAC Name: (2S,3S)-2-amino-3-methylpentanoic acid SMILES: CC[C@H](C)[C@H](N)C(O)=O
Isoleucine (L-Isoleucine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
L-Isoleucine, MedChemExpress
MedChemExpress L-isoleucine is a nonpolar hydrophobic amino acid. L-Isoleucine is an essential amino acid.
Isoleucine (L-Isoleucine), LoGiCal
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.