missing translation for 'onlineSavingsMsg'
Learn More

l-Isoleucine, Millipore™

Product Code. 10484741
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10484741 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ100 Graded - For Non-regulated Laboratory Applications
Product Code. 10484741 Supplier Millipore Supplier No. 1053620500
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ100 Graded - For Non-regulated Laboratory Applications

For biochemistry

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 279°C to 280°C (Decomposition)
CAS 73-32-5
MDL Number MFCD00064222
Synonym L-Isoleucine, iso-Leucine, Ile, L-Isoleucine
Molecular Weight (g/mol) 131.17
Quantity 500 g
Grade Research
Chemical Name or Material L-Isoleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.