accessibility menu, dialog, popup

UserName

NHOCl

CAS RN 69601-43-0

IUPAC Name: 4-chloro-N-cyclohexylbutanamide
Synonyms: 4-chloro-N-cyclohexylbutanamide
Molecular Weight (g/mol): 203.71
Molecular Formula: C10H18ClNO
InChi Key: KYKIYEJLXULPLV-UHFFFAOYSA-N
SMILES: ClCCCC(=O)NC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results